2-Fluoro-5-methoxy-3-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

86385-500MG 86385-1G 86385-5G
77334-920EA 310.03 USD
77334-920 77359-064 77383-004
2-Fluoro-5-methoxy-3-(methylthio)phenol ≥97%
2-Fluoro-5-methoxy-3-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₈H₉FO₂S
Storage Temperature: Ambient
CAS Number: 2710311-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR