3-Fluoro-2-hydroxy-6-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

86370-500MG 86370-1G 86370-5G
77337-646EA 433.63 USD
77337-646 77359-062 77383-002
3-Fluoro-2-hydroxy-6-methoxybenzaldehyde ≥97%
3-Fluoro-2-hydroxy-6-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520914
CAS Number: 170482-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR