(3-Fluoro-5-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

86284-500MG 86284-1G 86284-5G 86284-25G
77334-914EA 202.91 USD
77334-914 77359-060 77370-950 77391-512
(3-Fluoro-5-methylphenyl)trimethylsilane ≥95%
(3-Fluoro-5-methylphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₅FSi
MW: 182.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34687529
CAS Number: 1851745-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR