Trimethyl(2-methyl-4-(trifluoromethyl)phenyl)silane ≥95%

Supplier: AOBChem USA

Warning

86272-500MG 86272-1G 86272-5G 86272-25G
77334-912EA 263.68 USD
77334-912 77359-058 77370-948 77391-510
Trimethyl(2-methyl-4-(trifluoromethyl)phenyl)silane ≥95%
Trimethyl(2-methyl-4-(trifluoromethyl)phenyl)silane

Precaución: for Research Use Only

Formula: C₁₁H₁₅F₃Si
MW: 232.318 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770671
CAS Number: 1876969-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR