Trimethyl(3-(methylthio)phenyl)silane ≥97%

Supplier: AOBChem USA

Warning

86228-500MG 86228-1G 86228-5G 86228-25G
77334-908EA 244.11 USD
77334-908 77359-054 77370-944 77391-506
Trimethyl(3-(methylthio)phenyl)silane ≥97%
Trimethyl(3-(methylthio)phenyl)silane

Precaución: for Research Use Only

Formula: C₁₀H₁₆SSi
Storage Temperature: Ambient
MDL Number: MFCD34770660
CAS Number: 22515-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR