(3-Chloro-4-methoxyphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

86210-500MG 86210-1G 86210-5G 86210-25G
77334-906EA 202.91 USD
77334-906 77359-052 77370-942 77391-504
(3-Chloro-4-methoxyphenyl)trimethylsilane ≥97%
(3-Chloro-4-methoxyphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₅ClOSi
MW: 214.764 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30154794
CAS Number: 1864985-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR