Trimethyl(2-(trifluoromethoxy)phenyl)silane ≥97%

Supplier: AOBChem USA

Warning

86172-500MG 86172-1G 86172-5G 86172-25G
77334-902EA 243.08 USD
77334-902 77359-048 77370-938 77391-500
Trimethyl(2-(trifluoromethoxy)phenyl)silane ≥97%
Trimethyl(2-(trifluoromethoxy)phenyl)silane

Precaución: for Research Use Only

Formula: C₁₀H₁₃F₃OSi
MW: 234.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22370326
CAS Number: 1443345-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR