Trimethyl(3-(trifluoromethoxy)phenyl)silane ≥95%

Supplier: AOBChem USA

Warning

86163-500MG 86163-1G 86163-5G 86163-25G
77334-898EA 243.08 USD
77334-898 77359-044 77370-934 77391-496
Trimethyl(3-(trifluoromethoxy)phenyl)silane ≥95%
Trimethyl(3-(trifluoromethoxy)phenyl)silane

Precaución: for Research Use Only

Formula: C₁₀H₁₃F₃OSi
MW: 234.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22370327
CAS Number: 1443345-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR