Trimethyl(2,4,6-trifluorophenyl)silane ≥95%

Supplier: AOBChem USA

Warning

86157-500MG 86157-1G 86157-5G 86157-25G
77334-896EA 178.19 USD
77334-896 77346-706 77370-932 77391-494
Trimethyl(2,4,6-trifluorophenyl)silane ≥95%
Trimethyl(2,4,6-trifluorophenyl)silane

Precaución: for Research Use Only

Formula: C₉H₁₁F₃Si
Storage Temperature: Ambient
MDL Number: MFCD00092629
CAS Number: 51584-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR