(3,4-Dichlorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

86149-500MG 86149-1G 86149-5G 86149-25G
77334-892EA 130.81 USD
77334-892 77344-360 77370-928 77382-998
(3,4-Dichlorophenyl)trimethylsilane ≥97%
(3,4-Dichlorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₂Cl₂Si
Storage Temperature: Ambient
MDL Number: MFCD06201810
CAS Number: 149021-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR