(3-Fluoro-4-methylphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

86142-500MG 86142-1G 86142-5G 86142-25G
77334-890EA 220.42 USD
77334-890 77359-040 77370-926 77391-490
(3-Fluoro-4-methylphenyl)trimethylsilane ≥97%
(3-Fluoro-4-methylphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₅FSi
MW: 182.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201808
CAS Number: 845823-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR