(4-Fluoronaphthalen-1-yl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

86138-500MG 86138-1G 86138-5G 86138-25G
77334-888EA 220.42 USD
77334-888 77359-038 77370-924 77391-488
(4-Fluoronaphthalen-1-yl)trimethylsilane ≥97%
(4-Fluoronaphthalen-1-yl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₃H₁₅FSi
MW: 218.342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201821
CAS Number: 35869-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR