Trimethyl(2-(methylthio)phenyl)silane ≥97%

Supplier: AOBChem USA

Warning

86121-500MG 86121-1G 86121-5G 86121-25G
77334-886EA 243.08 USD
77334-886 77359-036 77370-922 77391-486
Trimethyl(2-(methylthio)phenyl)silane ≥97%
Trimethyl(2-(methylthio)phenyl)silane

Precaución: for Research Use Only

Formula: C₁₀H₁₆SSi
MW: 196.385 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617801
CAS Number: 33356-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR