Trimethyl(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)silane ≥97%

Supplier: AOBChem USA

Warning

86117-500MG 86117-1G 86117-5G
77338-964EA 536.63 USD
77338-964 77359-032 77382-996
Trimethyl(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)silane ≥97%
Trimethyl(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)silane

Precaución: for Research Use Only

Formula: C₁₅H₂₅BO₂Si
MW: 276.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383882
CAS Number: 1208070-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR