(5-Fluoro-2-methoxyphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

86113-500MG 86113-1G 86113-5G 86113-25G
77334-882EA 183.34 USD
77334-882 77347-144 77370-918 77391-482
(5-Fluoro-2-methoxyphenyl)trimethylsilane ≥97%
(5-Fluoro-2-methoxyphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₅FOSi
MW: 198.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11521265
CAS Number: 708-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR