2-Bromo-4-iodo-1,3-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

86111-500MG 86111-1G 86111-5G
77340-160EA 800.31 USD
77340-160 77359-030 77382-994
2-Bromo-4-iodo-1,3-dimethoxybenzene ≥97%
2-Bromo-4-iodo-1,3-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrIO₂
MW: 342.956 g/mol
Storage Temperature: Ambient
CAS Number: 1600352-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR