(3-Fluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

86094-500MG 86094-1G 86094-5G 86094-25G
77334-874EA 220.42 USD
77334-874 77359-026 77370-908 77391-476
(3-Fluorophenyl)trimethylsilane ≥97%
(3-Fluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₃FSi
MW: 168.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201839
CAS Number: 7217-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR