1-Bromo-4-(2,2-diethoxyethoxy)-3-fluoro-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

86064-500MG 86064-1G 86064-5G 86064-25G
77334-870EA 170.98 USD
77334-870 77346-386 77370-900 77391-472
1-Bromo-4-(2,2-diethoxyethoxy)-3-fluoro-2-methylbenzene ≥95%
1-Bromo-4-(2,2-diethoxyethoxy)-3-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrFO₃
MW: 321.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770615
CAS Number: 2901065-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR