5-Bromo-7-methylbenzofuran ≥97%

Supplier: AOBChem USA

Warning

86054-500MG 86054-1G 86054-5G 86054-25G
77334-866EA 145.23 USD
77334-866 77345-116 77370-896 77391-468
5-Bromo-7-methylbenzofuran ≥97%
5-Bromo-7-methylbenzofuran

Precaución: for Research Use Only

Formula: C₉H₇BrO
Storage Temperature: Ambient
MDL Number: MFCD11877829
CAS Number: 170681-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR