1-(4-Chloro-3-fluorophenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85895-500MG 85895-1G 85895-5G 85895-25G
77334-862EA 116.39 USD
77334-862 77343-474 77370-892 77382-982
1-(4-Chloro-3-fluorophenyl)cyclopentanol ≥95%
1-(4-Chloro-3-fluorophenyl)cyclopentanol

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClFO
MW: 214.664 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770551
CAS Number: 1075189-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR