3,5-Dibromo-4-fluoroaniline ≥97%

Supplier: AOBChem USA

Warning

85890-1G 85890-5G 85890-25G
77340-880EA 61.8 USD
77340-880 77370-890 77382-980
3,5-Dibromo-4-fluoroaniline ≥97%
3,5-Dibromo-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄Br₂FN
Storage Temperature: Ambient
MDL Number: MFCD09835195
CAS Number: 1003709-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR