1-(2-Isopropylphenyl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

85875-500MG 85875-1G 85875-5G 85875-25G
77334-860EA 184.37 USD
77334-860 77347-346 77370-888 77391-464
1-(2-Isopropylphenyl)cyclopentanol ≥97%
1-(2-Isopropylphenyl)cyclopentanol

Precaución: for Research Use Only

Formula: C₁₄H₂₀O
MW: 204.308 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770549
CAS Number: 65429-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR