1-(naphthalen-2-yl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85868-500MG 85868-1G 85868-5G 85868-25G
77334-858EA 136.99 USD
77334-858 77344-640 77370-886 77391-462
1-(naphthalen-2-yl)cyclopentanol ≥95%
1-(naphthalen-2-yl)cyclopentanol

Precaución: for Research Use Only

Formula: C₁₅H₁₆O
MW: 212.287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222249
CAS Number: 110971-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR