1-(2-Ethoxyphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85866-500MG 85866-1G 85866-5G 85866-25G
77334-856EA 182.31 USD
77334-856 77347-052 77370-884 77391-460
1-(2-Ethoxyphenyl)cyclopentanol ≥95%
1-(2-Ethoxyphenyl)cyclopentanol

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
MW: 206.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17223656
CAS Number: 1341976-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR