1-(2,6-Difluorophenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

85857-500MG 85857-1G 85857-5G 85857-25G
77334-854EA 87.55 USD
77334-854 77342-312 77370-882 77382-978
1-(2,6-Difluorophenyl)cyclopentanol ≥95%
1-(2,6-Difluorophenyl)cyclopentanol

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD19637737
CAS Number: 1342136-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR