1-(3,4-Dichlorophenyl)cyclobutanol ≥97%

Supplier: AOBChem USA

Warning

85776-500MG 85776-1G 85776-5G 85776-25G
77334-850EA 223.51 USD
77334-850 77359-018 77370-878 77391-456
1-(3,4-Dichlorophenyl)cyclobutanol ≥97%
1-(3,4-Dichlorophenyl)cyclobutanol

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD21788631
CAS Number: 1536771-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR