1-(3-(Trifluoromethyl)phenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

85729-500MG 85729-1G 85729-5G 85729-25G
77334-846EA 116.39 USD
77334-846 77343-472 77370-876 77382-974
1-(3-(Trifluoromethyl)phenyl)cyclobutanol ≥95%
1-(3-(Trifluoromethyl)phenyl)cyclobutanol

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O
Storage Temperature: Ambient
MDL Number: MFCD21788803
CAS Number: 164171-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR