2-Fluoro-1-(3-Fluoro-4-methoxyphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
85670-500MG
85670-1G
85670-5G
77339-622EA
603.58
USD
77339-622
77359-012
77382-972
2-Fluoro-1-(3-Fluoro-4-methoxyphenyl)ethanone ≥97%
2-Fluoro-1-(3-Fluoro-4-methoxyphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₈F₂O₂ MW: 186.155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13172691 CAS Number: 501426-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...