1-(2,4-Difluorophenyl)-2-fluoroethanone ≥97%

Supplier: AOBChem USA

Warning

85663-500MG 85663-1G 85663-5G
77337-770EA 440.84 USD
77337-770 77359-008 77382-968
1-(2,4-Difluorophenyl)-2-fluoroethanone ≥97%
1-(2,4-Difluorophenyl)-2-fluoroethanone

Precaución: for Research Use Only

Formula: C₈H₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD24692838
CAS Number: 319-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR