1-(5-Bromo-2-fluorophenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

85659-500MG 85659-1G 85659-5G 85659-25G
77334-836EA 263.68 USD
77334-836 77359-006 77370-868 77391-444
1-(5-Bromo-2-fluorophenyl)-2-fluoroethanone ≥95%
1-(5-Bromo-2-fluorophenyl)-2-fluoroethanone

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O
MW: 235.026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28674593
CAS Number: 1600511-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR