2-Fluoro-1-(2-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

85654-500MG 85654-1G 85654-5G
77334-834EA 342.99 USD
77334-834 77359-004 77382-966
2-Fluoro-1-(2-methoxyphenyl)ethanone ≥97%
2-Fluoro-1-(2-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24695407
CAS Number: 2967-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR