2-Fluoro-1-(p-tolyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

85641-500MG 85641-1G 85641-5G
77334-832EA 338.87 USD
77334-832 77358-998 77382-960
2-Fluoro-1-(p-tolyl)ethanone ≥95%
2-Fluoro-1-(p-tolyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉FO
MW: 152.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24688612
CAS Number: 37032-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR