1-(4-Bromophenyl)-2-fluoroethanone ≥97%

Supplier: AOBChem USA

Warning

85640-500MG 85640-1G 85640-5G
77335-458EA 353.29 USD
77335-458 77358-996 77382-958
1-(4-Bromophenyl)-2-fluoroethanone ≥97%
1-(4-Bromophenyl)-2-fluoroethanone

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MW: 217.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13173359
CAS Number: 403-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR