1-Bromo-2-chloro-4-fluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

85630-500MG 85630-1G 85630-5G
77334-830EA 346.08 USD
77334-830 77358-994 77382-956
1-Bromo-2-chloro-4-fluoro-5-iodobenzene ≥97%
1-Bromo-2-chloro-4-fluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFI
Storage Temperature: Ambient
MDL Number: MFCD32903680
CAS Number: 2460515-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR