4-Methyl-2-(phenylethynyl)aniline ≥95%

Supplier: AOBChem USA

Warning

85628-500MG 85628-1G
77340-458EA 1130.94 USD
77340-458 77358-992
4-Methyl-2-(phenylethynyl)aniline ≥95%
4-Methyl-2-(phenylethynyl)aniline

Precaución: for Research Use Only

Formula: C₁₅H₁₃N
Storage Temperature: Ambient
MDL Number: MFCD23141133
CAS Number: 13141-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR