4-Fluoro-2-(phenylethynyl)aniline ≥95%

Supplier: AOBChem USA

Warning

85627-500MG 85627-1G 85627-5G 85627-25G
77334-828EA 151.41 USD
77334-828 77345-412 77370-866 77391-442
4-Fluoro-2-(phenylethynyl)aniline ≥95%
4-Fluoro-2-(phenylethynyl)aniline

Precaución: for Research Use Only

Formula: C₁₄H₁₀FN
MW: 211.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24646152
CAS Number: 875930-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR