2-Fluoro-5-iodo-1-isopropyl-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

85357-500MG 85357-1G
77340-500EA 1309.13 USD
77340-500 77358-988
2-Fluoro-5-iodo-1-isopropyl-3-methylbenzene ≥95%
2-Fluoro-5-iodo-1-isopropyl-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂FI
MW: 278.105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770429
CAS Number: 2901085-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR