1-(2-Bromo-4,5-dimethoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

85342-500MG 85342-1G 85342-5G 85342-25G
77334-822EA 141.11 USD
77334-822 77344-944 77370-860 77391-436
1-(2-Bromo-4,5-dimethoxyphenyl)ethanone ≥97%
1-(2-Bromo-4,5-dimethoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
MW: 259.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00508283
CAS Number: 74746-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR