1,2-dimethoxy-4-(methylsulfonyl)benzene ≥95%

Supplier: AOBChem USA

Warning

85341-1G 85341-5G 85341-25G
77341-584EA 93.73 USD
77341-584 77370-858 77382-954
1,2-dimethoxy-4-(methylsulfonyl)benzene ≥95%
1,2-dimethoxy-4-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₂O₄S
MW: 216.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03789187
CAS Number: 33951-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR