(2-Bromo-4,5-dimethoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

85333-500MG 85333-1G 85333-5G 85333-25G
77334-820EA 267.8 USD
77334-820 77358-986 77370-856 77391-434
(2-Bromo-4,5-dimethoxyphenyl)(methyl)sulfane ≥97%
(2-Bromo-4,5-dimethoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂S
MW: 263.151 g/mol
Storage Temperature: Ambient
CAS Number: 142294-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR