2-Fluoro-1-iodo-4-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

85326-500MG 85326-1G 85326-5G 85326-25G
77334-818EA 84.46 USD
77334-818 77342-208 77370-854 77382-952
2-Fluoro-1-iodo-4-isopropoxybenzene ≥97%
2-Fluoro-1-iodo-4-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FIO
MW: 280.078 g/mol
Storage Temperature: Ambient
CAS Number: 1369948-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR