3-Methoxy-4'-(trifluoromethoxy)-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

85309-500MG 85309-1G 85309-5G
77334-814EA 348.14 USD
77334-814 77358-984 77382-950
3-Methoxy-4'-(trifluoromethoxy)-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3-Methoxy-4'-(trifluoromethoxy)-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃O₃
MW: 296.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770418
CAS Number: 1191053-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR