3-Methoxy-4'-(trifluoromethoxy)-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

85307-500MG 85307-1G 85307-5G
77338-112EA 464.53 USD
77338-112 77358-982 77382-948
3-Methoxy-4'-(trifluoromethoxy)-[1,1'-biphenyl]-4-carboxylic acid ≥97%
3-Methoxy-4'-(trifluoromethoxy)-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃O₄
MW: 312.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18322446
CAS Number: 1261462-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR