3,4'-dimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

85299-500MG 85299-1G 85299-5G 85299-25G
77334-808EA 160.68 USD
77334-808 77345-804 77370-848 77391-428
3,4'-dimethoxy-[1,1'-biphenyl]-4-carbaldehyde ≥95%
3,4'-dimethoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770413
CAS Number: 2901082-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR