3,4'-dimethoxy-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

85297-500MG 85297-1G 85297-5G 85297-25G
77334-806EA 204.97 USD
77334-806 77358-976 77370-846 77391-426
3,4'-dimethoxy-[1,1'-biphenyl]-4-carboxylic acid ≥97%
3,4'-dimethoxy-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₄
MW: 258.269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18319526
CAS Number: 167627-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR