(3-Chloro-2,6-dimethoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

85199-500MG 85199-1G
77340-528EA 1455.39 USD
77340-528 77358-972
(3-Chloro-2,6-dimethoxyphenyl)(methyl)sulfane ≥97%
(3-Chloro-2,6-dimethoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂S
Storage Temperature: Ambient
MDL Number: MFCD34770411
CAS Number: 2711425-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR