3-Chloro-2-fluoro-5-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

85155-500MG 85155-1G 85155-5G
77339-276EA 571.65 USD
77339-276 77358-964 77382-932
3-Chloro-2-fluoro-5-methoxypyridine ≥97%
3-Chloro-2-fluoro-5-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFNO
Storage Temperature: Ambient
MDL Number: MFCD16610375
CAS Number: 1227574-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR