2-Bromo-N-methoxy-N,5-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

85109-500MG 85109-1G 85109-5G 85109-25G
77334-800EA 182.31 USD
77334-800 77347-050 77370-836 77391-424
2-Bromo-N-methoxy-N,5-dimethylbenzamide ≥97%
2-Bromo-N-methoxy-N,5-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂
MW: 258.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847172
CAS Number: 1048108-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR