(3-Bromo-2-chloro-5-isopropylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

85073-500MG 85073-1G 85073-5G
77338-028EA 459.38 USD
77338-028 77358-962 77382-930
(3-Bromo-2-chloro-5-isopropylphenyl)(methyl)sulfane ≥97%
(3-Bromo-2-chloro-5-isopropylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrClS
MW: 279.624 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770345
CAS Number: 2901099-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR