(3-Bromo-2-chloro-5-(trifluoromethoxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

85071-500MG 85071-1G 85071-5G
77336-850EA 394.49 USD
77336-850 77358-960 77382-928
(3-Bromo-2-chloro-5-(trifluoromethoxy)phenyl)(methyl)sulfane ≥97%
(3-Bromo-2-chloro-5-(trifluoromethoxy)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₅BrClF₃OS
Storage Temperature: Ambient
MDL Number: MFCD34770343
CAS Number: 2167440-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR